Learn More
Biomatik Corporation Mouse Protein disulfide-isomerase(P4HB) ELISA kit
Supplier:  Biomatik Corporation EKC37657
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09103. Target Synonyms: DSI, ERBA2L, GIT, P4Hbeta, PDI, PDIA1, PHDB, PO4DB, PO4HB, PROHB, collagen prolyl 4-hydroxylase beta|glutathione-insulin transhydrogenase|procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4. Restriction: For Research Use Only.
Catalog No. 50-229-4461