Learn More
Biomatik Corporation Mouse R-spondin-3(RSPO3) ELISA kit
Supplier:  Biomatik Corporation EKC37755
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 4.7 pg/mL-300 pg/mL. Sensitivity: 1.17 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q2TJ95. Target Synonyms: CRISTIN1, FLJ14440, PWTSR, THSD2, R-spondin 3|thrombospondin, type I, domain containing 2|thrombospondin-like. Restriction: For Research Use Only.
Catalog No. 50-229-4466