Learn More
Biomatik Corporation Mouse soluble Lectin-like Oxidized Low Density Lipoprotein Receptor-1(sLOX-1)ELISA Kit
Supplier:  Biomatik Corporation EKC37837
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9EQ09. Target Synonyms: CLEC8A, LOX1, SCARE1, lectin-type oxidized LDL receptor 1|oxidised low density lipoprotein (lectin-like) receptor 1|scavenger receptor class E, member 1. Restriction: For Research Use Only.
Catalog No. 50-229-4472