Learn More
Biomatik Corporation Mouse Thyroid-Peroxidase, Tpo Elisa Kit
Supplier:  Biomatik Corporation EKC37925
Encompass_Preferred
Mouse Thyroid-Peroxidase, Tpo Elisa Kit for detecting Thyroid Peroxidase in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.5-40ng/ml. Sensitivity: 2.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P35419; Tpo. Target Synonyms: Tpo; Thyroid peroxidase; TPO; EC 1.11.1.8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4477