Learn More
Biomatik Corporation Mouse Vascular endothelial cell growth factor receptor 2,VEGFR-2/Flk-1 ELISA kit
Supplier:  Biomatik Corporation EKC38030
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35918. Target Synonyms: CD309, FLK1, VEGFR, VEGFR2, fetal liver kinase-1|kinase insert domain receptor|protein-tyrosine kinase receptor Flk-1|soluble VEGFR2|tyrosine kinase growth factor receptor|vascular endothelial growt. Restriction: For Research Use Only.
Catalog No. 50-229-4486