Learn More
Biomatik Corporation Pig coagulation factor VIII, FⅦI ELISA Kit
Supplier:  Biomatik Corporation EKC38136
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 62.5 ng/mL-4000 ng/mL. Sensitivity: 15.62 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P12263. Target Synonyms: RP11-115M6.7, AHF, DXS1253E, F8B, F8C, FVIII, HEMA, coagulation factor VIII|coagulation factor VIIIc|factor VIII F8B|coagulation factor 8,F8. Restriction: For Research Use Only.
Catalog No. 50-229-4495