Learn More
Biomatik Corporation Pig Colipase(CLPS) ELISA kit
Supplier:  Biomatik Corporation EKC38142
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 15.6 mU/mL-1000 mU/mL. Sensitivity: 3.9 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P02703. Target Synonyms: colipase|pancreatic colipase preproprotein. Restriction: For Research Use Only.
Catalog No. 50-229-4497