Learn More
Biomatik Corporation Pig creatine kinase MB isoenzyme (CK-MB) ELISA kit
Supplier:  Biomatik Corporation EKC38155
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 2.35 mU/mL-150 mU/mL. Sensitivity: 0.58 mU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4500