Learn More
Biomatik Corporation Rabbit Immunoglobulin A, Iga Elisa Kit
Supplier:  Biomatik Corporation EKC38454
Encompass_Preferred
Rabbit Immunoglobulin A, Iga Elisa Kit for detecting Immunoglobulin A, Iga in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 12-3000ng/ml. Sensitivity: 3ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4524