Learn More
Biomatik Corporation Rabbit Motilin (Mln) Elisa Kit
Supplier:  Biomatik Corporation EKC38498
Encompass_Preferred
Rabbit Motilin (Mln) Elisa Kit for detecting Motilin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P27114; MLN. Target Synonyms: MLN; Promotilin [Cleaved into: Motilin; Motilin-associated peptide; MAP)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4529