Learn More
Biomatik Corporation Rabbit oxidized low density lipoprotein,OxLDL ELISA Kit
Supplier:  Biomatik Corporation EKC38508
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.078 μmol/mL-5 μmol/mL. Sensitivity: 0.02 μmol/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4531