Learn More
Biomatik Corporation Rabbit Plasminogen Activator Inhibitor 1, Pai-1 Elisa Kit
Supplier:  Biomatik Corporation EKC38517
Encompass_Preferred
Rabbit Plasminogen Activator Inhibitor 1, Pai-1 Elisa Kit for detecting Plasminogen Activator Inhibitor 1, Pai-1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4533