Learn More
Biomatik Corporation Rat Alpha-synuclein(SNCA) ELISA kit
Supplier:  Biomatik Corporation EKC38644
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P37377. Target Synonyms: MGC110988, NACP, PARK1, PARK4, PD1, OTTHUMP00000161559|OTTHUMP00000161561|alpha-synuclein|alpha-synuclein, isoform NACP140|non A-beta component of AD amyloid. Restriction: For Research Use Only.
Catalog No. 50-229-4547