Learn More
Biomatik Corporation Rat Aquaporin 2,AQP-2 ELISA Kit
Supplier:  Biomatik Corporation EKC38706
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.25 ng/mL-400 ng/mL. Sensitivity: 1.56 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P34080. Target Synonyms: AQP-CD, MGC34501, WCH-CD, aquaporin 2|aquaporin-CD|collecting duct water channel protein|water-channel aquaporin 2. Restriction: For Research Use Only.
Catalog No. 50-229-4553