Learn More
Biomatik Corporation Rat Beta-Arrestin-2 (Arrb2) Elisa Kit
Supplier:  Biomatik Corporation EKC38738
Encompass_Preferred
Rat Beta-Arrestin-2 (Arrb2) Elisa Kit for detecting Arrestin, Beta 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.72pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29067; Arrb2. Target Synonyms: Arrb2Beta-arrestin-2; Arrestin beta-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4555