Learn More
Biomatik Corporation Rat bradykinin,BK ELISA Kit
Supplier:  Biomatik Corporation EKC38759
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.78 pg/mL-50 pg/mL. Sensitivity: 0.195 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4557