Learn More
Biomatik Corporation Rat Carboxypeptidase B(CPB1) ELISA kit
Supplier:  Biomatik Corporation EKC38786
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 12.5 ng/mL-800 ng/mL. Sensitivity: 3.12 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P19223. Target Synonyms: CPB, DKFZp779K1333, PASP, PCPB, pancreas-specific protein|pancreatic carboxypeptidase B1|procarboxypeptidase B. Restriction: For Research Use Only.
Catalog No. 50-229-4559