Learn More
Biomatik Corporation Rat Cathepsin L1 (Ctsl1) Elisa Kit
Supplier:  Biomatik Corporation EKC38808
Encompass_Preferred
Rat Cathepsin L1 (Ctsl1) Elisa Kit for detecting Cathepsin L1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07154; Ctsl. Target Synonyms: Ctsl; Ctsl1Cathepsin L1; EC 3.4.22.15; Cathepsin L; Cyclic protein 2; CP-2; Major excreted protein; MEP) [Cleaved into: Procathepsin L; Cathepsin L1 heavy chain; Cathepsin L1 light chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4561