Learn More
Biomatik Corporation Rat Cytochrome P450 19A1(CYP19A1) ELISA kit
Supplier:  Biomatik Corporation EKC38941
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 18.75 pg/mL-1200 pg/mL. Sensitivity: 4.68 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P22443. Target Synonyms: ARO, ARO1, CPV1, CYAR, CYP19, MGC104309, P-450AROM, OTTHUMP00000162543|aromatase|cytochrome P450, subfamily XIX (aromatization of androgens)|estrogen synthetase|flavoprotein-linked monooxygenase|mic. Restriction: For Research Use Only.
Catalog No. 50-229-4569