Learn More
Biomatik Corporation Rat Epidermal growth factor,EGF ELISA Kit
Supplier:  Biomatik Corporation EKC38998
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07522. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4571