Learn More
Biomatik Corporation Rat Estradiol Receptor, Er Elisa Kit
Supplier:  Biomatik Corporation EKC39009
Encompass_Preferred
Rat Estradiol Receptor, Er Elisa Kit for detecting Estrogen Receptor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06211; Esr1. Target Synonyms: Esr1; Esr; Estr; Nr3a1; Estrogen receptor; ER; ER-alpha; Estradiol receptor; Nuclear receptor subfamily 3 group A member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4573