Learn More
Biomatik Corporation Rat Extracellular Superoxide Dismutase [Cu-Zn] (Cu/Zn-Sod/Sod3) Elisa Kit
Supplier:  Biomatik Corporation EKC39015
Encompass_Preferred
Rat Extracellular Superoxide Dismutase [Cu-Zn] (Cu/Zn-Sod/Sod3) Elisa Kit for detecting Superoxide Dismutase 3, Extracellular in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q08420; Sod3. Target Synonyms: Sod3; Sod-3; Extracellular superoxide dismutase [Cu-Zn]; EC-SOD; EC 1.15.1.1; Superoxide dismutase B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4575