Learn More
Biomatik Corporation Rat Fibronectin type III domain-containing protein 5(FNDC5) ELISA kit
Supplier:  Biomatik Corporation EKC39049
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8K3V5. Target Synonyms: FRCP2. Restriction: For Research Use Only.
Catalog No. 50-229-4578