Learn More
Biomatik Corporation Rat Glucocorticoid Receptor, Gr Elisa Kit
Supplier:  Biomatik Corporation EKC39103
Encompass_Preferred
Rat Glucocorticoid Receptor, Gr Elisa Kit for detecting Nuclear Receptor Subfamily 3, Group C, Member 1 (Glucocorticoid Receptor) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06536; Nr3c1. Target Synonyms: Nr3c1; GrlGlucocorticoid receptor; GR; Nuclear receptor subfamily 3 group C member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4581