Learn More
Biomatik Corporation Rat Indoleamine 2, 3-Dioxygenase, Ido Elisa Kit
Supplier:  Biomatik Corporation EKC39218
Encompass_Preferred
Rat Indoleamine 2, 3-Dioxygenase, Ido Elisa Kit for detecting Indoleamine 2, 3-Dioxygenase 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9ERD9; Ido1. Target Synonyms: Ido1; Ido; Indo; Indoleamine 2; 3-dioxygenase 1; IDO-1; EC 1.13.11.52; Indoleamine-pyrrole 2; 3-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4593