Learn More
Biomatik Corporation Rat interferon-inducible protein 10,IP-10 ELISA Kit
Supplier:  Biomatik Corporation EKC39249
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 781.25 pg/mL-50000 pg/mL. Sensitivity: 195 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48973. Target Synonyms: C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1, gamma IP10|interferon-inducible cytokine IP-10|protein 10 from interferon (gamma)-induced cell line|small inducible cytokine B10|small inducibl. Restriction: For Research Use Only.
Catalog No. 50-229-4595