Learn More
Biomatik Corporation Rat Interleukin 16,IL-16 ELISA KIT
Supplier:  Biomatik Corporation EKC39258
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: G5BEZ3. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4597