Learn More
Biomatik Corporation Rat Neurotensin (Nts) Elisa Kit
Supplier:  Biomatik Corporation EKC39483
Encompass_Preferred
Rat Neurotensin (Nts) Elisa Kit for detecting Neurotensin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20068; Nts. Target Synonyms: NtsNeurotensin/neuromedin N [Cleaved into: Large neuromedin N; NmN-125); Neuromedin N; NN; NmN); Neurotensin; NT); Tail peptide]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4616