Learn More
Biomatik Corporation Rat Osteopontin,OPN ELISA Kit
Supplier:  Biomatik Corporation EKC39514
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.06 ng/ml -50 ng/ml. Sensitivity: 0.06 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08721. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4618