Learn More
Biomatik Corporation Rat Protein S100-A6 (S100A6) Elisa Kit
Supplier:  Biomatik Corporation EKC39612
Encompass_Preferred
Rat Protein S100-A6 (S100A6) Elisa Kit for detecting S100 Calcium Binding Protein A6 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.4-150ng/ml. Sensitivity: 0.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P30801. Target Synonyms: S100A6; Protein S100-A6; Calcyclin; Lung 10 kDa protein; S100 calcium-binding protein A6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4632