Learn More
Biomatik Corporation Rat Sodium/Potassium-Transporting Atpase Subunit Alpha-1 (Atp1A1) Elisa Kit
Supplier:  Biomatik Corporation EKC39697
Encompass_Preferred
Rat Sodium/Potassium-Transporting Atpase Subunit Alpha-1 (Atp1A1) Elisa Kit for detecting Atpase, Na+/K+ Transporting, Alpha 1 Polypeptide in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06685; Atp1a1. Target Synonyms: Atp1a1Sodium/potassium-transporting ATPase subunit alpha-1; Na (+)/K (+) ATPase alpha-1 subunit; EC 7.2.2.13; Sodium pump subunit alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4642