Learn More
Biomatik Corporation Rat Troponin I, slow skeletal muscle(TNNI1) ELISA kit
Supplier:  Biomatik Corporation EKC39823
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P13413. Target Synonyms: DKFZp451O223, SSTNI, TNN1, OTTHUMP00000033877|OTTHUMP00000033910|slow-twitch skeletal troponin I|troponin I, skeletal, slow. Restriction: For Research Use Only.
Catalog No. 50-229-4656