Learn More
Biomatik Corporation Sheep Prolactin/Luteotropic Hormone(PRL/LTH) ELISA Kit
Supplier:  Biomatik Corporation EKC39984
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Ovis aries (Sheep). Detection Range: 5 μIU/mL-2000 μIU/mL. Sensitivity: 5 μIU/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01240. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4672