Learn More
Biomatik Corporation Mouse Angiotensin converting enzyme,ACE ELISA Kit
Supplier:  Biomatik Corporation EKC40026
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09470. Target Synonyms: ACE1, CD143, DCP, DCP1, MGC26566, MVCD3, CD143 antigen|angiotensin I converting enzyme 1|angiotensin converting enzyme, somatic isoform|carboxycathepsin|dipeptidyl carboxypeptidase 1|kininase II|pep. Restriction: For Research Use Only.
Catalog No. 50-229-4677