Learn More
Biomatik Corporation Human secondary lymphoid-tissue chemokine,SLC ELISA Kit
Supplier:  Biomatik Corporation EKC40033
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O00585. Target Synonyms: 6Ckine, CKb9, ECL, MGC34555, SCYA21, SLC, TCA4, Efficient Chemoattractant for Lymphocytes|beta chemokine exodus-2|exodus-2|secondary lymphoid tissue chemokine|small inducible cytokine A21|small indu. Restriction: For Research Use Only.
Catalog No. 50-229-4679