Learn More
Biomatik Corporation Mouse E-Selectin ELISA kit
Supplier:  Biomatik Corporation EKC40038
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q00690. Target Synonyms: RP1-117P20.2, CD62E, ELAM, ELAM1, ESEL, LECAM2, endothelial adhesion molecule 1|leukocyte endothelial cell adhesion molecule 2. Restriction: For Research Use Only.
Catalog No. 50-229-4682