Learn More
Biomatik Corporation Mouse Interleukin 16,IL-16 ELISA KIT
Supplier:  Biomatik Corporation EKC40073
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O54824. Target Synonyms: FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, LCF, prIL-16, OTTHUMP00000190849|OTTHUMP00000190850|interleukin 16|lymphocyte chemoattractant factor. Restriction: For Research Use Only.
Catalog No. 50-229-4693