Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein-3Α, Mip-3Α Elisa Kit
Supplier:  Biomatik Corporation EKC40098
Encompass_Preferred
Human Macrophage Inflammatory Protein-3Α, Mip-3Α Elisa Kit for detecting Chemokine (C-C Motif) Ligand 20 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78556; CCL20. Target Synonyms: Beta chemokine exodus 1; Beta-chemokine exodus-1; C C motif chemokine ligand 20; C-C motif chemokine 20; CC chemokine LARC; Ccl20; CCL20 (2-70); CCL20_HUMAN; Chemokine (C C motif) ligand 20; Chemokine C-C motif chemokine 20; Chemokine CC motif ligand 20; CKb4; Exodus 1; Exodus; LARC; Liver and activation regulated chemokine; Liver and activation-regulated chemokine; Macrophage inflammatory protein 3 alpha; MIP 3 alpha; MIP 3A; MIP-3-alpha; MIP-3a; MIP3 alpha; MIP3A; SCYA20; Small inducible cytokine A20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4695