Learn More
Biomatik Corporation Human Alpha-Fetoprotein, Afp Elisa Kit
Supplier:  Biomatik Corporation EKC40134
Encompass_Preferred
Human Alpha-Fetoprotein, Afp Elisa Kit for detecting Alpha-Fetoprotein in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 1.332ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02771; AFP. Target Synonyms: Afp; AFPD; Alpha fetoglobulin; Alpha fetoprotein; Alpha fetoprotein precursor; Alpha-1-fetoprotein; Alpha-fetoglobulin; Alpha-fetoprotein; alpha-fetoprotein; Hereditary persistence of; included; FETA; FETA_HUMAN; Hereditary persistence of alpha fetoprotein; HPAFP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4707