Learn More
Biomatik Corporation Rat Interferon β,IFN-β/IFNB ELISA Kit
Supplier:  Biomatik Corporation EKC40151
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P70499. Target Synonyms: IFB, IFF, IFNB, MGC96956, interferon beta. Restriction: For Research Use Only.
Catalog No. 50-229-4709