Learn More
Biomatik Corporation Rabbit basic fibroblast growth factor,bFGF ELISA Kit
Supplier:  Biomatik Corporation EKC40258
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48799. Target Synonyms: BFGF, FGFB, HBGF-2, basic fibroblast growth factor bFGF|fibroblast growth factor 2|heparin-binding growth factor 2|prostatropin. Restriction: For Research Use Only.
Catalog No. 50-229-4719