Learn More
Biomatik Corporation Rat transforming growth factor α,TGF-α ELISA Kit
Supplier:  Biomatik Corporation EKC40278
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8 pg/ml-500pg/ml. Sensitivity: 1.95 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01134. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4721