Learn More
Biomatik Corporation Rat Connective Tissue Growth Factor, Ctgf Elisa Kit
Supplier:  Biomatik Corporation EKC40339
Encompass_Preferred
Rat Connective Tissue Growth Factor, Ctgf Elisa Kit for detecting Connective Tissue Growth Factor in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9R1E9; Ctgf. Target Synonyms: Ccn2; CtgfCCN family member 2; Cellular communication network factor 2; Connective tissue growth factor; Connective tissue growth-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4741