Learn More
Biomatik Corporation Rat Neuron-specific enolase,NSE ELISA Kit
Supplier:  Biomatik Corporation EKC40355
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07323. Target Synonyms: NSE, 2-phospho-D-glycerate hydrolyase|enolase 2|neural enolase|neuron specific gamma enolase|neurone-specific enolase. Restriction: For Research Use Only.
Catalog No. 50-229-4745