Learn More
Biomatik Corporation Human Ceruloplasmin, Cp/Cer Elisa Kit
Supplier:  Biomatik Corporation EKC40434
Encompass_Preferred
Human Ceruloplasmin, Cp/Cer Elisa Kit for detecting Ceruloplasmin (Ferroxidase) in serum, plasma, cell culture supernates, urine, saliva. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00450; CP. Target Synonyms: CERU_HUMAN; Ceruloplasmin; CP 2; CP; CP2; Ferroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4754