Learn More
Biomatik Corporation Human Angiotensin I, Ang-I Elisa Kit
Supplier:  Biomatik Corporation EKC40463
Encompass_Preferred
Human Angiotensin I, Ang-I Elisa Kit for detecting Angiotensin I, Ang-I in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: angiotensin I; Ang-I; angiotensin 1; Ang-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4756