Learn More
Biomatik Corporation Dog Renin Elisa Kit
Supplier:  Biomatik Corporation EKC40499
Encompass_Preferred
Dog Renin Elisa Kit for detecting Renin in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6DYE7; REN. Target Synonyms: REN; Renin; EC 3.4.23.15; Angiotensinogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4759