Learn More
Biomatik Corporation Rat visfatin ELISA Kit
Supplier:  Biomatik Corporation EKC40543
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q80Z29. Target Synonyms: 1110035O14Rik, DKFZp666B131, MGC117256, PBEF, PBEF1, VF, VISFATIN, NAmPRTase|pre-B cell-enhancing factor|pre-B-cell colony enhancing factor 1. Restriction: For Research Use Only.
Catalog No. 50-229-4767