Learn More
Biomatik Corporation Human Vitronectin (Vtn) Elisa Kit
Supplier:  Biomatik Corporation EKC40550
Encompass_Preferred
Human Vitronectin (Vtn) Elisa Kit for detecting Vitronectin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04004; VTN. Target Synonyms: Complement S Protein; Epibolin; S Protein; S-protein; Serum Spreading Factor; Serum-spreading factor; Somatomedin B; Somatomedin-B; V75; Vitronectin; Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN; VTNC_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4770