Learn More
Biomatik Corporation Pig Interleukin 1Α , Il-1Α Elisa Kit
Supplier:  Biomatik Corporation EKC40677
Encompass_Preferred
Pig Interleukin 1Α , Il-1Α Elisa Kit for detecting Interleukin 1, Alpha in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18430; IL1A. Target Synonyms: IL1A; Interleukin-1 alpha; IL-1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4777